Learn More
Crescent Chemical Co Inc PHTHALALDEHYDE ANALYTICAL GRDE
SDP
Supplier:  Crescent Chemical Co Inc SE32800.03
Encompass_Preferred
Phthalaldehyde analytical grade Especially purified for fluorimetric histidine determination (1). Reagent for amines and alkaloids (2) as well for amino acids (3, 4) and peptides (5, 6). Size - 100G Storage Conditions - +2 °C TO +8 °C Catalog Numb
Catalog No. 50-248-021