Learn More
Medchemexpress LLC Phthalazine | 253-52-1 | 130.15 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00201610G
Encompass_Preferred
Phthalazine is a nitrogen-containing organic heterocyclic compound that serves as a substrate for human aldehyde oxidase 1, which leads to the production of reactive oxygen species (ROS) and subsequent enzyme inactivation. It has demonstrated antitumor activity in various cancers, including blood cancer, breast cancer, colon cancer, lung cancer, and renal cancer.
  • Substrate for human aldehyde oxidase 1
  • Can lead to ROS production and enzyme inactivation
  • Nitrogen-containing organic heterocyclic compound
  • Exhibits antitumor activity in multiple cancer types
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Appears as a yellow to brown solid
Catalog No. 50-004-46425