Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phthalocyanine | 574-93-6 | 98.0% | 1 G
SDP

Supplier:  Medchemexpress LLC HY1213101G

Encompass_Preferred

Phthalocyanine is a macrocyclic organic photosensitizer and dye supplied as a solid for research use. It is employed in photodynamic studies, antibacterial photoinactivation research, pigments, and materials science applications.

  • Photosensitizer for photodynamic and antibacterial research.
  • Solid form suitable for storage and formulation.
  • Stable macrocyclic structure; CAS 574-93-6; molecular weight 514.54 g/mol.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month) under inert atmosphere.
  • Available in small research pack quantities.

Catalog No. 50-002-33743


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.