Learn More
Medchemexpress LLC Physalin F | 57423-71-9 | 10 MG

Supplier: Medchemexpress LLC HYN769610MG
Physalin F is a secosteroid with potent anti-inflammatory and immunomodulatory activities. It induces apoptosis of PBMC, decreasing the spontaneous proliferation and cytokine production caused by Human T-lymphotropic virus type 1 (HTLV-1) infection. Physalin F is a seco-steroid isolated from Physalis angulata L.
- Potent anti-inflammatory and immunomodulatory activities
- Induces apoptosis of PBMC
- Decreases spontaneous proliferation and cytokine production caused by Human T-lymphotropic virus type 1 (HTLV-1) infection
- High purity: 99.98%
- Molecular formula: C28H30O10
- Molecular weight: 526.53
- Appearance: White to off-white solid
- Soluble in DMSO at 100 mg/mL
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.