Learn More
Medchemexpress LLC Physalin F | 57423-71-9 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN7696
Encompass_Preferred
Physalin F is a secosteroid with potent anti-inflammatory and immunomodulatory activities. It induces apoptosis of PBMC, decreasing the spontaneous proliferation and cytokine production caused by Human T-lymphotropic virus type 1 (HTLV-1) infection.
  • Potent anti-inflammatory activity
  • Potent immunomodulatory activity
  • Induces apoptosis of PBMC
  • Decreases spontaneous proliferation and cytokine production caused by HTLV-1 infection
Catalog No. 50-004-20569