Learn More
Medchemexpress LLC Physalin F | 57423-71-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN76965MG
Encompass_Preferred
Physalin F is a secosteroid with potent anti-inflammatory and immunomodulatory activities. It induces apoptosis of peripheral blood mononuclear cells (PBMC), leading to a decrease in the spontaneous proliferation and cytokine production caused by Human T-lymphotropic virus type 1 (HTLV-1) infection.
  • Potent anti-inflammatory and immunomodulatory activities
  • Induces apoptosis of PBMC
  • Decreases spontaneous proliferation and cytokine production in HTLV-1 infection
  • High purity: 99.98%
Catalog No. 50-004-19658