Learn More
Medchemexpress LLC Physalin F | 57423-71-9 | 50 MG

Supplier: Medchemexpress LLC HYN769650MG
Physalin F is a secosteroid with potent anti-inflammatory and immunomodulatory activities. It induces apoptosis of PBMC, decreasing the spontaneous proliferation and cytokine production caused by Human T-lymphotropic virus type 1 (HTLV-1) infection. This product is intended for research use only and is not sold to patients. It is a seco-steroid derived from Physalis angulata L.
- Purity: 99.98%
- Molecular weight: 526.53
- Formula: C28H30O10
- Appearance: Solid
- Color: White to off-white
- Solubility (in vitro): DMSO: 100 mg/mL (189.92 mM; requires ultrasonic; hygroscopic DMSO significantly impacts solubility, use newly opened DMSO)
- Shipping: Room temperature in continental US
- Storage: 4°C, sealed storage, away from moisture and light
- Storage (in solvent): -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.