Learn More
Medchemexpress LLC Physcion-d3 (parietin-d3) | 1215751-27-1 | 99.8% | 287.28 | C16H9D3O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0108S10MG
Encompass_Preferred
Physcion-d3 (parietin-d3) is the deuterium-labeled isotopologue of physcion used as an analytical reference and tracer in biochemical research. It is supplied as a high-purity powder intended for laboratory use and mass-spectrometry quantification.
  • Deuterium-labeled internal standard for mass spectrometry
  • High chemical and isotopic purity (>99.8%)
  • Provided as a 10 mg powder for analytical workflows
  • Suitable for tracer and quantification studies
  • Stable when stored under recommended conditions
Catalog No. 50-002-95792