Learn More
AdipoGen PHYSIOLOGICAL SALINE 1.5 ML
SDP
Supplier:  AdipoGen IAX900003LD15
Encompass_Preferred
Physiological Saline [Sodium Chloride 0.9%] Endotoxin-free (sterile), AdipoGen Life Sciences. Chemical. MW: . Endotoxin free diluent buffer for dilution of S-type and R-type TLRpure LPS. For solubilisation of endotoxin-free reagents (e.g. ODNs) use PBS Endotoxin-free (sterile) 1.5ml (Cat. No.: IAX-900-001-LD15) or 10ml (Cat. No.: IAX-900-001-L010) or ddWater Endotoxin-free (sterile) 1.5ml (Cat. No.: IAX-900-002-LD15) or 10ml (Cat. No.: IAX-900-002-L010).
Catalog No. 50-596-731