Learn More
Cayman Chemical PhysostIgmInsalIcylate 50mg
SDP
Supplier:  Cayman Chemical 3310650 MG
Encompass_Preferred
An alkaloid and cholinergic agent; inhibits AChE (IC50 = 0.67 nM); inhibits ChE and increases vagal stimulation-induced ACh output in isolated chicken heart at 10 µM; induces decentralized lower eyelid contractions and increases blood pressure in anesthetized rats at 200 and 400 UG/kg
Catalog No. 50-295-0941