Learn More
Sigma Aldrich Fine Chemicals Biosciences Phytagel, 71010-52-1, MFCD00131909, 250 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P8169250G
Encompass
Phytagel is intended for use in place of agar and other gelling agent for plant tissue culture. Phytagel has been shown to be a superior substitute for tissue-culture-grade agar in the micropropagation of banana plantlets. BioReagent, plant cell culture tested, powder, Synonym: Agar substitute gelling agent, Gellan Gum.
Catalog No. NC0335270