Learn More
Sigma Aldrich Fine Chemicals Biosciences PhytatrayTM II L x W x H 1
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P5929250EA
Encompass
These trays are sterile disposable plant cell culture vessels made of PETG. They are ideal for numerous plant cell culture applications such as high throughput screening.
Catalog No. 50-290-4742