Learn More
TARGETMOL CHEMICALS INC PHYTIC ACID 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T297150MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Phytic acid (Fytic Acid) is a complexing agent for removal of traces of heavy metal ions. It acts also as a hypocalcemic agent. purity: 99%
Catalog No. 50-257-7026