Learn More
Cayman Chemical PhytnIc AcId 10mg
SDP
Supplier:  Cayman Chemical 9036010 MG
Encompass_Preferred
Phytanic acid is a saturated 20-carbon branched-chain fatty acid which can only be derived from dietary sources. In cases of peroxisomal disorders, elevated levels of phytanic acid have been observed, and this is linked to diseases such as infantile phytanic acid storage disease and Refsum’s disease.{1912}
Catalog No. 50-289-4898