Learn More
CPC Scientific H-gamma-Glu-Cys-gamma-Glu-Cys-gamma-Glu-Cys-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PHYT004A
Encompass
A glutathione-derived heavy metal-detoxifying peptide of higher plants consisting of 3 units of gammaGlu-Cys.SEQUENCE: H-gamma-Glu-Cys-gamma-Glu-Cys-gamma-Glu-Cys-Gly-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: (gammaE-C)3-GMOLECULAR FORMULA: C26H41N7O14S3MOLECULAR WEIGHT: 771.83STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: Cadystin A, Cadystin Protein, S Pombe (gamma-Glu-Cys)3-Gly, H-(gamma-Glu-Cys)3-Gly-OHRESEARCH AREA: MiscellaneousREFERENCES:Grill, E. et al. Science 230, 674 (1985) Rauser, WE. Plant Physiol. 109, 1141 (1995)SKU(s): PHYT-004A
Catalog No. 50-211-0152