Learn More
Medchemexpress LLC Phytochelatin 5 (PC 5) | 99465-97-1 | 99.6% | 1236.35 | 10 MG

Supplier: Medchemexpress LLC HYP332210MG
Phytochelatin 5 is a metal-binding compound produced by plants. It may reduce the bioavailability of dietary toxic metals such as cadmium. This compound is for research use only and not sold to patients.
- Appearance: Solid
- Color: White to off-white
- Formula: C42H65N11O22S5
- Storage: Sealed, away from moisture (powder: -80°C for 2 years, -20°C for 1 year; in solvent: -80°C for 6 months, -20°C for 1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.