Learn More
Medchemexpress LLC Phytochelatin 5 (PC 5) | 99465-97-1 | 99.6% | 1236.35 | 5 MG

Supplier: Medchemexpress LLC HYP33225MG
Phytochelatin 5 is a metal-binding compound produced by plants that may reduce the bioavailability of dietary toxic metals such as cadmium.
- Metal-binding compound
- Produced by plants
- Reduces bioavailability of dietary toxic metals such as cadmium
- Molecular weight: 1236.35
- Formula: C42H65N11O22S5
- CAS No.: 99465-97-1
- Appearance: Solid
- Color: White to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.