Learn More
Medchemexpress LLC Phytochelatin 5 (PC 5) | 99465-97-1 | 99.62% | 1236.35 | 100 MG

Supplier: Medchemexpress LLC HYP3322100MG
Phytochelatin 5 is a metal-binding compound produced by plants. It may reduce the bioavailability of dietary toxic metals such as cadmium. This biochemical assay reagent appears as a white to off-white solid.
- Metal-binding compound
- Produced by plants
- Reduces bioavailability of dietary toxic metals such as cadmium
- Appears as a white to off-white solid
- Suitable for biochemical assays
- Store powder at -80°C for 2 years or -20°C for 1 year
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.