Learn More
Medchemexpress LLC Phytochelatin 5 (PC 5) | 99465-97-1 | 99.62% | 1236.35 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP3322
Phytochelatin 5 is a metal-binding compound produced by plants. It may reduce the bioavailability of dietary toxic metals such as cadmium. This product is a white to off-white solid with a purity of 99.62%.
- Reduces the bioavailability of dietary toxic metals
- Purity of 99.62%
- White to off-white solid appearance
- Recommended storage: sealed and away from moisture
- Powder stable for up to 2 years at -80°C
- In solvent, stable for up to 6 months at -80°C
- Soluble in DMSO at 25 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.