Learn More
Medchemexpress LLC Phytohemagglutinin P | 9008-97-3 | MFCD00165279 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN70385MG
Encompass_Preferred
Phytohemagglutinin P (PHA-P) is a mitogen that selectively stimulates cells of hematogenous or lymphoid monocytic origin. It is intended for research use only, supporting studies on cellular responses and proliferation.
  • Selectively stimulates cells of hematogenous or lymphoid monocytic origin
  • Stimulates decidual CD8+ T lymphocytes to produce interferon gamma and interleukin 8
  • Appears as a white to light yellow solid
  • Derived from plants (Leguminosae, Glycine max (L.) merr)
Catalog No. 50-004-18955