Learn More
Medchemexpress LLC Phytol | 150-86-7 | 1 ML
Supplier: Medchemexpress LLC HYN307510MM/1ML
Phytol ((E)-Phytol) is an orally active diterpenoid alcohol extracted from chlorophyll. It exhibits antioxidant, anti-inflammatory, anti-schistosomiasis, and antibacterial activities. It is intended for research use only and not for sale to patients.
- Exhibits antioxidant activity
- Shows anti-inflammatory properties
- Possesses anti-schistosomiasis activity
- Demonstrates antibacterial activities
- Reduces motor activity and causes death in worms
- Inhibits osteoclast differentiation and oxidative stress
- Reduces parasite numbers in mice infected with C. mansoni
- Improves anemia and oxidative brain damage in Plasmodium berghei infected mice