Learn More
Medchemexpress LLC Phytolaccagenin | 1802-12-6 | 532.71 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN14335MG
Encompass_Preferred
Phytolaccagenin, a triterpenoid saponin, is the active component of Radix Phytolaccae. It exhibits antifungal activity, anti-inflammatory activity, and lower toxicity.
  • Purity: 99.86%
  • Active component: Triterpenoid saponin from Radix Phytolaccae
  • Biological activities:
    • Antifungal activity
    • Anti-inflammatory activity
    • Lower toxicity
  • For research use only
Catalog No. 50-004-15991