Learn More
Cayman Chemical PhytosphngosInyeast 5mg
SDP
Supplier:  Cayman Chemical 202175 MG
Encompass_Preferred
A sphingolipid; metabolized to odd-numbered fatty acids with 2-hydroxy palmitic acid as an intermediate; induces cell death in vitro; inhibits carbachol-induced PLD activation in CHO cells transfected with C. elegans muscarinic acetylcholine receptors; essential in the yeast heat stress response
Catalog No. 50-298-2952