Learn More
TARGETMOL CHEMICALS INC PI-103 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T614325MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PI-103 is a potent cell-permeable ATP-competitive inhibitor of PI3K family members (IC50s 2/3/3/15/30/23 nM for p110(alpha)/(beta)/(delta)/(gamma) mTOR and DNA-PK). purity: 99%
Catalog No. 50-257-6592