Learn More
Medchemexpress LLC PI-103 hydrochloride | 371935-79-4 | MFCD10687102 | ≥98.0% | 384.82 g/mol | C19H17ClN4O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10115A10MM 1ML
Encompass_Preferred
PI-103 hydrochloride is a small-molecule research reagent that functions as a dual inhibitor of PI3K and mTOR. Supplied as a hydrochloride salt, it is used in cellular and biochemical studies to probe PI3K/mTOR signaling pathways and can be purchased as a solid or as a pre-made 10 mM solution in DMSO for immediate use.
  • Dual PI3K and mTOR inhibition for pathway studies.
  • Available as a 10 mM solution in DMSO for convenient use.
  • Also available as a solid for custom formulation.
  • High purity suitable for research (≥98%).
  • Applicable to cellular and biochemical assay workflows.
Catalog No. 50-002-11459