Learn More
Echelon Biosciences Research Labs PI(3,4)P2 PIP Beads 1 mL
SDP
Supplier:  Echelon Biosciences Research Labs PB034A1ML
Encompass
PI(3,4)P2 PIP BeadsTM are composed of agarose with 10 nanomoles of bound PI(3,4)P2 per mL of beads, enough for several protein binding experiment. Each 1 mL order of PIP Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
Catalog No. 50-720-685