Learn More
Echelon Biosciences Research Labs PI(3 4)P2 MASS ELISA
SDP
Supplier:  Echelon Biosciences Research Labs K3800EAKIT
Encompass
The PI(3,4)P2 Mass ELISA measures the amount of PI(3,4)P2 extracted from cells by means of a competitive ELISA (96 well format).Assay Range: 300 to 1.23 pmol PI(3,4)P2Sample Type: Dried down lipid extracted from cellsSample Volume: 120 L/sample, run in duplicate. This item is non returnable
Catalog No. 50-720-450