Learn More
Echelon Biosciences Research Labs PI(5)P DIC8 AMMONIUM SALT 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs P5008A100UG
Encompass
Phosphatidylinositol 5-phosphate diC8 (PI(5)P diC8) is a synthetic, purified dioctanoyl PI(5)P.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 5-phosphate (PI(5)P) is a rare lipid found in the nucleus that can bind the PHD finger of the nuclear adapter protein ING2. PI(5)P is phosphorylated to PI(4,5)P2 by Type II PIP kinases. This item is non returnable
Catalog No. 50-720-645