Learn More
Novus Biologicals PI MARKER 10.10
SDP
Supplier:  Novus Biologicals 102232
Encompass
Marker for pI determination that will give you both consistent and accurate measurement of isoelectric point. Can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample protein's isoelectric point, and for quality control of sample analysis. You'll get 200 aL of pI marker stock solution in every vial. 10.10 pI value.
Catalog No. NC0432904