Learn More
Novus Biologicals PI MARKER 5.85
SDP
Supplier:  Novus Biologicals 102225
Encompass
APC/Cy7 anti-mouse/rat CD29; Clone: HMbeta1-1; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC
Catalog No. NC9548707