Learn More
Novus Biologicals iCE280/iCE3 pI Marker 6.14, 200ul
SDP
Supplier:  Novus Biologicals 102220
Encompass
We have a wide range of markers for pI determination that will give you both consistent and accurate measurement of isoelectric point on your iCE system. They can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample protein's isoelectric point, and for quality control of sample analysis. You'll get 200 L of pI marker stock solution in every vial.
Catalog No. NC0285361