Learn More
Novus Biologicals PI MARKER, pI Value 7.4
SDP
Supplier:  Novus Biologicals 102227
Encompass
Markers for pI determination that will give you both consistent and accurate measurement of isoelectric point on your iCE system. They can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample protein's isoelectric point, and for quality control of sample analysis. You'll get 200 µL of pI marker stock solution in every vial.
Catalog No. NC9603833