Learn More
Novus Biologicals PI MARKER 8.4
SDP
Supplier:  Novus Biologicals 102229
Encompass
Marker for pI determination that will give you both consistent and accurate measurement of isoelectric point on your iCE system. They can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample protein's isoelectric point, and for quality control of sample analysis. You'll get 200 L of pI marker stock solution in every vial.
Catalog No. NC9518931