Learn More
Novus Biologicals iCE280/iCE3 pI Marker 8.79, 200ul
SDP
Supplier:  Novus Biologicals 102230
Encompass
pI Markers, pI Value 8.79 *Markers for pI determination that will give you both consistent and accurate measurement of isoelectric point on your iCE system. *Can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample protein's isoelectric point, and for quality control of sample analysis. *You'll get 200 L of pI marker stock solution in every vial.
Catalog No. NC9673175