Learn More
Novus Biologicals PI MARKER PI 9.50 0.4MG
SDP
Supplier:  Novus Biologicals 101996
Encompass
Can be used for whole-column and single-point capillary isoelectric focusing (cIEF) detection to measure your sample's isoelectric point. 200uL of pI marker stock solution in every vial. This product's pI value is 9.50.
Catalog No. NC9103783