Learn More
Cayman Chemical LY294002, 154447-36-6, 10 mg
SDP
Supplier:  Cayman Chemical 7092010
Encompass_Preferred
Molecular Formula C19H17NO3, Purity ≥98%, Formula Weight 307.3; A selective PI3K inhibitor with an IC50 value of 1.4 M.
Catalog No. NC9363690