Learn More
Selleck Chemical LLC PI3K/AKT-IN-1 E4683-1G
SDP
Supplier:  Selleck Chemical LLC E46831G
Encompass_Preferred
PI3K/AKT-IN-1 is a dual-acting inhibitor of PI3K/AKT pathway with an IC50 of 6 99 M 4 01 M and 3 36 M for PI3K PI3K and AKT-1 respectively It also induces S-phase cell cycle arrest caspase-3-dependent apoptosis exhibiting strong potential against breast cancer and leukemia
Catalog No. 50-000-39095