Learn More
Medchemexpress LLC PI3K-IN-2 | 1403458-28-5 | 99.6% | 525.54 g/mol | C28H29F2N3O5 | 5 MG

Supplier: Medchemexpress LLC HY1015175MG
PI3K-IN-2 is a selective, orally active small-molecule inhibitor of PI3Kβ and PI3Kδ used in preclinical research. It exhibits low-nanomolar potency and favorable selectivity versus other PI3K isoforms, and is supplied as a high-purity solid suitable for biochemical and cellular studies.
- Potent PI3Kβ/δ inhibition (low-nanomolar activity).
- Selectivity versus PI3Kσ and PI3Kγ.
- High purity (~99.6%).
- Molecular weight 525.54 g/mol and formula C28H29F2N3O5.
- Available in multiple milligram quantities for research use.
- Stable as a powder at -20°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.