Learn More
Medchemexpress LLC PI3Kα-IN-4 | 2322293-83-2 | 99.8% | C25H23ClFN5O5S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13134510MG
Encompass_Preferred
PI3Kα-IN-4 is a selective, orally active small-molecule inhibitor of PI3Kα with potent biochemical activity (IC50 1.8 nM) and demonstrated antitumor efficacy in vivo. Supplied as an off-white to yellow solid with high purity, it is formulated for research use with characterized solubility and storage profiles suitable for in vitro and in vivo studies.
  • Selective PI3Kα inhibition with potent biochemical potency (IC50 1.8 nM).
  • Demonstrated cellular activity and antitumor efficacy in preclinical models.
  • High chemical purity suitable for research applications.
  • Good solubility in DMSO for preparation of concentrated stock solutions.
  • Stable solid form with defined refrigerated and frozen storage recommendations.
Catalog No. 50-002-60875