Learn More
Medchemexpress LLC PI3K/mTOR Inhibitor-13 (sodium) | 2361009-23-4 | C20H13F2N5NaO3S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY153120A50MG
Encompass_Preferred
PI3K/mTOR Inhibitor-13 (sodium) is an orally active dual inhibitor of phosphoinositol 3-kinase (PI3K) and mTOR kinase. It shows potential applications in sexual diseases, solid tumor, and idiopathic pulmonary fibrosis (IPF). Intended for research use only, this white to off-white solid has a purity of 98.11% and a molecular weight of 464.40.
  • Inhibits HFL1 cell proliferation in a dose-dependent manner.
  • Increases α-SMA/Tubulin expression in HFL1 cells.
  • Evaluated for pharmacokinetic parameters in rats, mice, dogs, and monkeys.
Catalog No. 50-003-73039