Learn More
Medchemexpress LLC PI3K/mTOR Inhibitor-2 | 1848242-58-9 | 99.1% | 478.86 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11150850MG
Encompass_Preferred
PI3K/mTOR Inhibitor-2 is a potent dual pan-PI3K/mTOR inhibitor. It exhibits IC50s of 3.4 nM for PI3Kα, 34 nM for PI3Kβ, 16 nM for PI3Kδ, 1 nM for PI3Kγ, and 4.7 nM for mTOR. The product also demonstrates antitumor activity. For research use only.
  • Potent dual pan-PI3K/mTOR inhibitor
  • Exhibits antitumor activity
  • IC50 for PI3Kα: 3.4 nM
  • IC50 for PI3Kβ: 34 nM
  • IC50 for PI3Kδ: 16 nM
  • IC50 for PI3Kγ: 1 nM
  • IC50 for mTOR: 4.7 nM
Catalog No. 50-003-45999