Learn More
TARGETMOL CHEMICALS INC PI3KALPHA/MTOR-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T125895MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. PI3K(alpha)/mTOR-IN-1 is a potent dual inhibitor of PI3K(alpha)/mTOR. PI3K(alpha)/mTOR-IN-1 shows an IC50 of 7 nM for PI3K(alpha) in a cell assay and Kis of 12.5 nM and 10.6 nM for mTOR and PI3K(alpha) in a cell free assay respectively. purity: 99%
Catalog No. 50-258-1438