Learn More
ABclonal Technology PIBF1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A12033500UL
Encompass_Preferred
This gene encodes a protein that is induced by the steroid hormone progesterone and plays a role in the maintenance of pregnancy. The encoded protein regulates multiple facets of the immune system to promote normal pregnancy including cytokine synthesis, natural killer (NK) cell activity, and arachidonic acid metabolism. Low serum levels of this protein have been associated with spontaneous pre-term labor in humans. This protein may promote the proliferation, migration and invasion of glioma.
Catalog No. 50-000-98913