Learn More
Medchemexpress LLC Picalm protein, human (HEK293, His) | >95.0% | 50UG
SDP
Supplier:  Medchemexpress LLC HYP70113550UG
Encompass_Preferred
Recombinant human PICALM protein produced in HEK293 cells with a C-terminal His tag, supplied as a 50 μg vial for research use. The protein is >95% pure by reducing SDS-PAGE and has an approximate molecular weight of 90-150 kDa. Store long-term at -80°C and avoid repeated freeze-thaw cycles. Intended applications include biochemical assays, antibody production, and protein-protein interaction studies.
  • High purity suitable for sensitive assays.
  • HEK293 expression ensures mammalian post-translational modifications.
  • C-terminal His tag enables straightforward purification and detection.
  • Supplied in small aliquots to minimize freeze-thaw damage.
  • Includes supporting documents (SDS, COA, data sheet) for quality verification.
Catalog No. 50-001-76965