Learn More
Medchemexpress LLC Picamilon sodium | 62936-56-5 | 97.0% | 230.20 | C10H11N2NaO3 | 1 G
SDP
Supplier:  Medchemexpress LLC HY107482A1G
Encompass_Preferred
Picamilon sodium is the monosodium salt of nicotinoyl-γ-aminobutyric acid (GABA), provided as a research-grade, water-soluble powder for biochemical and pharmacological studies. The compound is typically supplied at research-scale amounts and is characterized by a molecular weight of 230.20 g/mol and a typical purity of 97.0%.
  • Monosodium salt of nicotinoyl-GABA for research use.
  • Orally active GABA derivative suitable for pharmacological studies.
  • Water-soluble powder for easy formulation and dosing.
  • High purity (approximately 97.0%) for reproducible results.
  • Molecular weight 230.20 g/mol; formula C10H11N2NaO3.
Catalog No. 50-002-41251