Learn More
Medchemexpress LLC Picaridin 500mg | 119515-38-7 | 500 MG

Supplier: Medchemexpress LLC HY116144500MG
Picaridin (Icaridin) is a research-grade insect repellent and analytical reference compound used in olfactory, repellency, and antimicrobial studies. It is supplied in solid and solution forms for laboratory use and intended for research and analytical applications only (not for human use).
- CAS number 119515-38-7.
- Chemical formula C12H23NO3.
- Molecular weight 229.32 g/mol.
- Purity reported around 99% (see lot-specific COA for exact value).
- Available in 500 mg and larger gram-scale quantities, and as prepared solutions (for example, 10 mM in DMSO).
- Suitable as an analytical standard and for repellency and olfactory testing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.