Learn More
AdipoGen Piceatannol (25 mg)
SDP
Supplier:  AdipoGen AGCN20086M025
Encompass_Preferred
Chemical. CAS: 10083-24-6. Formula: C14H12O4. MW: 244.3. Synthetic. Originally isolated from Euphorbia lagascae. Wide range of tyrosine and serine/threonine protein kinase inhibitor, including Syk, p56lck, PKA, PKC, MLCK, CDPK, JNK and PI3K. Inhibits the tyrosine phosphorylation of STAT3 and STAT5. Potent apoptosis inducer. Potent anticancer compound. Suppresses NF-kB activation through IkBalpha kinase inhibition. Activator of human deacetylase SIRT1 (sirtuin 1). Potent antioxidant with anti-proliferative, anti-inflammatory and cardioprotective properties. Neuoprotective. Adipogenesis inhibitor. Promotes glucose uptake, AMPK phosphorylation and GLUT4 translocation. Autophagy inducer
Catalog No. 50-205-3445