Learn More
Medchemexpress LLC HY-N2237 1mg Medchemexpress, Piceatannol 3'-O-glucoside CAS:94356-26-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22371MG
Encompass_Preferred
Medchemexpress, HY-N2237 1mg Piceatannol 3'-O-glucoside CAS:94356-26-0 Piceatannol 3'-O-glucoside, an active component of Rhubarb, activates endothelial nitric oxide (NO) synthase through inhibition of arginase activity with IC50s of 11.22 µM and 11.06 µM against arginase I and arginase II, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2880