Learn More
Research Products International Corp Picloram (4-Amino-3,5,6-Trichloropicolinic Acid), 10 Grams
SDP
Supplier:  Research Products International Corp P4012010.0
Encompass_Preferred
Auxin like growth regulator.
Catalog No. 50-213-618