Learn More
Medchemexpress LLC HY-101507 5mg Medchemexpress, Pico145 CAS:1628287-16-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1015075MG
Encompass_Preferred
Medchemexpress, HY-101507 5mg Pico145 CAS:1628287-16-0 Pico145 (HC-608) is a remarkable inhibitor of TRPC1/4/5 channels, inhibits (−)-englerin A-activated TRPC4/TRPC5 channels, with IC50s of 0.349 and 1.3 nM in cells, and shows no effect on TRPC3, TRPC6, TRPV1, TRPV4, TRPA1, TRPM2, TRPM8. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8662